Order Entry
Ireland
ContactUsLinkComponent
Human COL6A3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human COL6A3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5808-96
Supplier:  ANTIBODIES.COM
New Product
Human COL6A3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human COL6A3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5808-96
Supplier:  ANTIBODIES.COM