Order Entry
Ireland
ContactUsLinkComponent
Mouse INSL6 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse INSL6 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
  ANTIA5598-96
New Product
Mouse INSL6 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse INSL6 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
  ANTIA5598-96