Order Entry
Ireland
ContactUsLinkComponent
Bovine MMP7 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of bovine MMP7 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3941-96
Supplier:  ANTIBODIES.COM
New Product
Bovine MMP7 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of bovine MMP7 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3941-96
Supplier:  ANTIBODIES.COM