Order Entry
Ireland
ContactUsLinkComponent
Porcine Orexin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of porcine Orexin in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA82290-96
Supplier:  ANTIBODIES.COM
New Product
Porcine Orexin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of porcine Orexin in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA82290-96
Supplier:  ANTIBODIES.COM