Human Urocanase Domain Containing Protein 1/UROC1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for th e in vitro quantitative determination of human Urocanase Domain Containing Protein 1/UROC1 in tissue homogenates, cel 1 * 96 Tests
Catalog # ANTIA8164-96
Supplier: ANTIBODIES.COM