Order Entry
Ireland
ContactUsLinkComponent
Rat CYP11B1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat CYP11B1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6499-48
Supplier:  ANTIBODIES.COM
New Product
Rat CYP11B1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat CYP11B1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6499-48
Supplier:  ANTIBODIES.COM