Order Entry
Ireland
ContactUsLinkComponent
Human SLC12A3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human SLC12A3 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6495-48
Supplier:  ANTIBODIES.COM
New Product
Human SLC12A3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human SLC12A3 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6495-48
Supplier:  ANTIBODIES.COM