Order Entry
Ireland
ContactUsLinkComponent
Human Nrf2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human Nrf2 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA7064-96
New Product
Human Nrf2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human Nrf2 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA7064-96