Order Entry
Ireland
ContactUsLinkComponent
Rabbit PPAR gamma ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of rabbit PPAR gamma in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 96 Tests
Catalog # ANTIA7718-96
Supplier:  ANTIBODIES.COM
New Product
Rabbit PPAR gamma ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of rabbit PPAR gamma in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 96 Tests
Catalog # ANTIA7718-96
Supplier:  ANTIBODIES.COM