Order Entry
Ireland
ContactUsLinkComponent
Human Cubilin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human Cubilin in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6636-96
Supplier:  ANTIBODIES.COM
New Product
Human Cubilin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human Cubilin in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6636-96
Supplier:  ANTIBODIES.COM