Order Entry
Ireland
ContactUsLinkComponent
Human IRS1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human IRS1 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA82428-96
Supplier:  ANTIBODIES.COM
New Product
Human IRS1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human IRS1 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA82428-96
Supplier:  ANTIBODIES.COM