Order Entry
Ireland
ContactUsLinkComponent
Human Testis Anion Transporter 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human Testis Anion Transporter 1 in tissue homogenates, cell lysates or other biological flui 1 * 48 Tests
Catalog # ANTIA7270-48
Supplier:  ANTIBODIES.COM
New Product
Human Testis Anion Transporter 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human Testis Anion Transporter 1 in tissue homogenates, cell lysates or other biological flui 1 * 48 Tests
Catalog # ANTIA7270-48
Supplier:  ANTIBODIES.COM