Order Entry
Ireland
ContactUsLinkComponent
Human RAMP2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human RAMP2 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6044-96
Supplier:  ANTIBODIES.COM
New Product
Human RAMP2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human RAMP2 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6044-96
Supplier:  ANTIBODIES.COM