Order Entry
Ireland
ContactUsLinkComponent
Human Complement C4a ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Complement C4a in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA82300-96
Supplier:  ANTIBODIES.COM
New Product
Human Complement C4a ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Complement C4a in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA82300-96
Supplier:  ANTIBODIES.COM