Order Entry
Ireland
ContactUsLinkComponent
Rat IL-11RA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat IL-11RA in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7348-96
Supplier:  ANTIBODIES.COM
New Product
Rat IL-11RA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat IL-11RA in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7348-96
Supplier:  ANTIBODIES.COM