Order Entry
Ireland
ContactUsLinkComponent
Human FUT3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human FUT3 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6608-96
Supplier:  ANTIBODIES.COM
New Product
Human FUT3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human FUT3 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6608-96
Supplier:  ANTIBODIES.COM