Order Entry
Ireland
ContactUsLinkComponent
Human IL-21R ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IL-21R in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7820-48
Supplier:  ANTIBODIES.COM
New Product
Human IL-21R ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IL-21R in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7820-48
Supplier:  ANTIBODIES.COM