Order Entry
Ireland
ContactUsLinkComponent
Rat ZO1 Tight Junction Protein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of rat ZO1 Tight Junction Protein in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA8046-48
Supplier:  ANTIBODIES.COM
New Product
Rat ZO1 Tight Junction Protein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of rat ZO1 Tight Junction Protein in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA8046-48
Supplier:  ANTIBODIES.COM