Order Entry
Ireland
ContactUsLinkComponent
Human AMN ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human AMN in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA6631-96
New Product
Human AMN ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human AMN in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA6631-96