Order Entry
Ireland
ContactUsLinkComponent
Human RBM3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human RBM3 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA8138-48
Supplier:  ANTIBODIES.COM
New Product
Human RBM3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human RBM3 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA8138-48
Supplier:  ANTIBODIES.COM