Order Entry
Ireland
ContactUsLinkComponent
Human Cdc25A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Cdc25A in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7344-96
Supplier:  ANTIBODIES.COM
New Product
Human Cdc25A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Cdc25A in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7344-96
Supplier:  ANTIBODIES.COM