Order Entry
Ireland
ContactUsLinkComponent
Rat MYH8 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat MYH8 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6306-48
Supplier:  ANTIBODIES.COM
New Product
Rat MYH8 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat MYH8 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6306-48
Supplier:  ANTIBODIES.COM