Order Entry
Ireland
ContactUsLinkComponent
Rat Presenilin 2/AD5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat Presenilin 2/AD5 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7243-96
Supplier:  Antibodies.com
New Product
Rat Presenilin 2/AD5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat Presenilin 2/AD5 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7243-96
Supplier:  Antibodies.com