Order Entry
Ireland
ContactUsLinkComponent
Mouse CELA2A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse CELA2A in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7973-96
Supplier:  ANTIBODIES.COM
New Product
Mouse CELA2A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse CELA2A in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7973-96
Supplier:  ANTIBODIES.COM