Order Entry
Ireland
ContactUsLinkComponent
Mouse LGALS3BP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse LGALS3BP in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6387-48
Supplier:  ANTIBODIES.COM
New Product
Mouse LGALS3BP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse LGALS3BP in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6387-48
Supplier:  ANTIBODIES.COM