Order Entry
Ireland
ContactUsLinkComponent
Bovine S100A9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of bovine S100A9 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6313-96
Supplier:  ANTIBODIES.COM
New Product
Bovine S100A9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of bovine S100A9 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6313-96
Supplier:  ANTIBODIES.COM