Order Entry
Ireland
ContactUsLinkComponent
Mouse MTAP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse MTAP in tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA7807-48
New Product
Mouse MTAP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse MTAP in tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA7807-48