Order Entry
Ireland
ContactUsLinkComponent
Human JAK3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human JAK3 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA8060-96
New Product
Human JAK3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human JAK3 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA8060-96