Order Entry
Ireland
ContactUsLinkComponent
Bovine CXCL11 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of bovine CXCL11 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6314-48
Supplier:  ANTIBODIES.COM
New Product
Bovine CXCL11 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of bovine CXCL11 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6314-48
Supplier:  ANTIBODIES.COM