Order Entry
Ireland
ContactUsLinkComponent
Human S100 Calcium Binding Protein A13/S100A13 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the i n vitro quantitative determination of human S100 Calcium Binding Protein A13/S100A13 in serum, plasma, tissue homogen 1 * 96 Tests
Catalog # ANTIA74527-96
Supplier:  ANTIBODIES.COM
New Product
Human S100 Calcium Binding Protein A13/S100A13 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the i n vitro quantitative determination of human S100 Calcium Binding Protein A13/S100A13 in serum, plasma, tissue homogen 1 * 96 Tests
Catalog # ANTIA74527-96
Supplier:  ANTIBODIES.COM