Order Entry
Ireland
ContactUsLinkComponent
Human CYP11B1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human CYP11B1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA82308-96
Supplier:  ANTIBODIES.COM
New Product
Human CYP11B1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human CYP11B1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA82308-96
Supplier:  ANTIBODIES.COM