Order Entry
Ireland
ContactUsLinkComponent
Mouse Histone H2B (Testis Specific) ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of mouse Histone H2B (Testis Specific) in serum, plasma, tissue homogenates, and other biolog 1 * 96 Tests
Catalog # ANTIA74000-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Histone H2B (Testis Specific) ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of mouse Histone H2B (Testis Specific) in serum, plasma, tissue homogenates, and other biolog 1 * 96 Tests
Catalog # ANTIA74000-96
Supplier:  ANTIBODIES.COM