Order Entry
Ireland
ContactUsLinkComponent
Human ESAM ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human ESAM in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA6367-96
New Product
Human ESAM ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human ESAM in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA6367-96