Order Entry
Ireland
ContactUsLinkComponent
Mouse BCMA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse BCMA in serum, plasma or other biological fluids. 1 * 96 Tests
  ANTIA7759-96
New Product
Mouse BCMA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse BCMA in serum, plasma or other biological fluids. 1 * 96 Tests
  ANTIA7759-96