Order Entry
Ireland
ContactUsLinkComponent
Mouse DPP9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse DPP9 in tissue homogenates or other biological fluids. 1 * 96 Tests
  ANTIA6158-96
New Product
Mouse DPP9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse DPP9 in tissue homogenates or other biological fluids. 1 * 96 Tests
  ANTIA6158-96