Order Entry
Ireland
ContactUsLinkComponent
Human G-Protein Coupled Receptor 30 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of human G-Protein Coupled Receptor 30 in serum, plasma, tissue homogenates or other biologic 1 * 96 Tests
Catalog # ANTIA6409-96
Supplier:  ANTIBODIES.COM
New Product
Human G-Protein Coupled Receptor 30 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of human G-Protein Coupled Receptor 30 in serum, plasma, tissue homogenates or other biologic 1 * 96 Tests
Catalog # ANTIA6409-96
Supplier:  ANTIBODIES.COM