Order Entry
Ireland
ContactUsLinkComponent
Mouse G-6-Pase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse G-6-Pase in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7801-96
Supplier:  ANTIBODIES.COM
New Product
Mouse G-6-Pase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse G-6-Pase in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7801-96
Supplier:  ANTIBODIES.COM