ND-Rückschlagsucgerzbg-Acetylen/oxyge
Catalog # MSPP1500-O
Supplier: MARKET SOURCE PART PROCESS
New Product
ND-Rückschlagsucgerzbg-Acetylen/oxyge
Catalog # MSPP1500-O
Supplier: MARKET SOURCE PART PROCESS
New Product
Frequently Bought Together
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Diethyl ether, EMPARTA® ACS for analysis, Supelco®
Supplier: Merck
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
4-Methyl-2-pentanone ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Petroleum spirit, 60…95 °C, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: Merck
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals