Order Entry
Ireland
ContactUsLinkComponent
Potassium Permanganate 0.002M (0.01N) CRM, ISO 17034 and ISO 17025,,,, Conservation 12 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C,, 1 * 1 L
Catalog # ACSDZ02564114.1L
Supplier:  CPACHEM
New Product
Potassium Permanganate 0.002M (0.01N) CRM, ISO 17034 and ISO 17025,,,, Conservation 12 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C,, 1 * 1 L
Catalog # ACSDZ02564114.1L
Supplier:  CPACHEM