Order Entry
Ireland
ContactUsLinkComponent
 
TOYOPEARL® Super A Affinity Chromatography Protein Resins
  TOYO0023581
New Product
 :  
TOYOPEARL® Super A Affinity Chromatography Protein Resins
  TOYO0023581
New Product
 :  

 

  • Particle Size:
    45 µm
  • Pore size:
    100 nm
  • Description:
    TOYOPEARL Super A protein resin
  • Seq:
    2
  • pH range:
    3 - 12 (operational); 2 - 14 (CIP)
  • Matrix:
    Hydroxylated methacrylic polymer
  • Ligand:
    Modified, code-optimised C domain, hexameric ligand with multipoint attachment
  • Binding capacity:
    72 mg mAb/ml resin @ 5 min RT; 60 mg mAb/ml resin @ 3 min RT
  • Concentration:
    Endotoxin concentration: Max 10 EU/ml
  • Flow rate:
    10 - 600 cm/h (linear)
  • Formulation:
    72% slurry in 20% ethanol
  • Volume:
    25 ml

 

 

TOYOPEARL® Super A resin provides a cutting-edge experience in protein A media, blending top-tier critical performance attributes with exceptional operational flexibility. An efficient and durable protein A resin.

  • State-of-the-art operational flexibility

    Supports high titers with high dynamic binding capacity (DBC)

    Enhanced alkaline stability for rigorous CIP

    Excellent pressure flow behavior to increase process efficiency

    Superior critical process performance

    Features mild elution conditions

    Reduction in process impurities, makes process robust, safe, and compliant

An enhanced, animal-origin free, recombinant protein A ligand is bound to the polymethacrylate base bead via multipoint attachment resulting in excellent base stability for up to 100 CIP cycles with 0,5 mol/L NaOH and up to 70 CIP cycles with 1 mol/L NaOH while maintaining 90% of its initial DBC.

TOYOPEARL Super A resin is available in ~45 µm particle size (F-grade) and exhibits dynamic binding capacities of greater than 70 g/L at 5 min residence time and greater than 60 g/L at 3 min residence time.

Improved mass transfer characteristics allow it to maintain a larger percent of its capacity at lower residence times relative to comparable agarose-based resins. It is therefore ideally suited for continuous operation using multiple short columns.

This resin is also available in pre-packed Skill Pak columns to simplify your process development and scale-up.

Bacterial count: max 100 CFU/ml

Endotoxin concentration: max 10 EU/ml

Delivery conditions: 72% slurry in 20% ethanol

Frequently Bought Together