Order Entry
Ireland
ContactUsLinkComponent
Nylon Syringe Filters, 10.0um, 25mm, Luer-Lock/Luer Slip, Nonsterile, 100 per pack, SF18126 1 * 100 items
Catalog # MSPPSF18126
Nylon Syringe Filters, 10.0um, 25mm, Luer-Lock/Luer Slip, Nonsterile, 100 per pack, SF18126 1 * 100 items
Catalog # MSPPSF18126
Frequently Bought Together