Order Entry
Ireland
ContactUsLinkComponent
Phosphonic acid CAS:13598-36-2 EC:237-066-7 CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store in a refrigerator at temperatures between 2C to 8C 1 * 250 mg
Catalog # ACSDSB38810.250MG
Supplier:  CPACHEM
New Product
Phosphonic acid CAS:13598-36-2 EC:237-066-7 CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store in a refrigerator at temperatures between 2C to 8C 1 * 250 mg
Catalog # ACSDSB38810.250MG
Supplier:  CPACHEM