Stainless steel tray, non-perforated, 15 mm rim, may affect the temperature distribution, for in 1 * 1 items
MEMMB02073IN160
: MEMMERT
New Product
Stainless steel tray, non-perforated, 15 mm rim, may affect the temperature distribution, for in 1 * 1 items
MEMMB02073IN160
: MEMMERT
New Product
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection