Order Entry
Ireland
ContactUsLinkComponent
Comfort MTP rack for upright freezers for SBS format (HxD) 3x7=21 plates, 90mm high , stainless steel, dimensions (HxDxW) 277x630x13 5mm 1 * 1 items
Catalog # TENATE35680
Supplier:  TENAK
New Product
Comfort MTP rack for upright freezers for SBS format (HxD) 3x7=21 plates, 90mm high , stainless steel, dimensions (HxDxW) 277x630x13 5mm 1 * 1 items
Catalog # TENATE35680
Supplier:  TENAK