Order Entry
Ireland
ContactUsLinkComponent
Comfort MTP rack for upright freezers for SBS format (HxD) 2x5=10 plates, 90mm high , stainless steel, dimensions (HxDxW) 185x450x13 5mm 1 * 1 items
Catalog # TENATE35666
Supplier:  TENAK
New Product
Comfort MTP rack for upright freezers for SBS format (HxD) 2x5=10 plates, 90mm high , stainless steel, dimensions (HxDxW) 185x450x13 5mm 1 * 1 items
Catalog # TENATE35666
Supplier:  TENAK