Order Entry
Ireland
ContactUsLinkComponent
Comfort MTP rack for upright freezers for SBS format (HxD) 11x5=55 plates, 35mm high , stainless steel, dimensions (HxDxW) 408x450x1 35mm 1 * 1 items
Catalog # TENATE35322
Supplier:  TENAK
New Product
Comfort MTP rack for upright freezers for SBS format (HxD) 11x5=55 plates, 35mm high , stainless steel, dimensions (HxDxW) 408x450x1 35mm 1 * 1 items
Catalog # TENATE35322
Supplier:  TENAK