Order Entry
Ireland
ContactUsLinkComponent
Standard rack for upright freezers for (HxD) 8x4=32 TENAK Microscope slide boxes (87x128mm), 32mm high, stainless steel, dimensions (HxDxW) 289x363x135mm 1 * 1 items
Catalog # TENATE30126
Supplier:  TENAK
New Product
Standard rack for upright freezers for (HxD) 8x4=32 TENAK Microscope slide boxes (87x128mm), 32mm high, stainless steel, dimensions (HxDxW) 289x363x135mm 1 * 1 items
Catalog # TENATE30126
Supplier:  TENAK