Order Entry
Ireland
ContactUsLinkComponent
Standard MTP rack for upright freezers for SBS format (HxD) 8x7=56 plates, 35mm high, stainless steel, dimensions (HxDxW) 297x630x13 5mm 1 * 1 items
Catalog # TENATE27340
Supplier:  TENAK
New Product
Standard MTP rack for upright freezers for SBS format (HxD) 8x7=56 plates, 35mm high, stainless steel, dimensions (HxDxW) 297x630x13 5mm 1 * 1 items
Catalog # TENATE27340
Supplier:  TENAK