Order Entry
Ireland
ContactUsLinkComponent
Standard MTP rack for upright freezers for SBS format (HxD) 10x6=60 plates, 35mm high, stainless steel, dimensions (HxDxW) 371x540x1 35mm 1 * 1 items
Catalog # TENATE27332
Supplier:  TENAK
New Product
Standard MTP rack for upright freezers for SBS format (HxD) 10x6=60 plates, 35mm high, stainless steel, dimensions (HxDxW) 371x540x1 35mm 1 * 1 items
Catalog # TENATE27332
Supplier:  TENAK