Order Entry
Ireland
ContactUsLinkComponent
Comfort rack for chest freezers for 9 cryoboxes, 75mm high, stainless steel, with spring lock, dimensions (HxDxW) 715x140x140mm, del ivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24544B10
Supplier:  TENAK
New Product
Comfort rack for chest freezers for 9 cryoboxes, 75mm high, stainless steel, with spring lock, dimensions (HxDxW) 715x140x140mm, del ivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24544B10
Supplier:  TENAK